CAS RN 7240-90-6
5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside, 98+%
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 MDL Number: MFCD00005666 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure, Thermo Scientific Chemicals
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.63 MDL Number: MFCD00005666 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
Thermo Scientific Chemicals 5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99+%, for molecular biology, DNAse, RNAse and Protease free
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.63 MDL Number: MFCD00005666 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, MP Biomedicals™
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
5-Bromo-4-Chloro-3-Indolyl-β-D-Galactopyranoside, ≥98%, Molecular Biology Reagent, MP Biomedicals™
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 MDL Number: MFCD00005666 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, MP Biomedicals™
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 MDL Number: MFCD00005666 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
5-Bromo-4-chloro-3-indolyl beta-D-Galactopyranoside 98.0+%, TCI America™
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 MDL Number: MFCD00005666 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
5-Bromo-4-chloro-3-indolyl-βD-galactopyranoside, Promega
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
Fisher Chemical X-Gal, 5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, Fisher BioReagents™
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
MilliporeSigma™ X-Gal, OmniPur™, 100mg, Calbiochem™,
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br
Avantor Macron X-Gal, Macron Fine Chemicals
CAS: 7240-90-6 Molecular Formula: C14H15BrClNO6 Molecular Weight (g/mol): 408.629 InChI Key: OPIFSICVWOWJMJ-AEOCFKNESA-N Synonym: x-gal,5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole,5-bromo-4-chloro-3-indolyl-beta-d-galactoside,unii-v595og374w,5-bromo-4-chloro-3-indolyl beta-galactoside,5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactoside,5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside,5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside,indoxyl-gal PubChem CID: 65181 ChEBI: CHEBI:75055 IUPAC Name: (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br