Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ X-Gal, OmniPur™, 100mg, Calbiochem™,

Catalog No. 9630100MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
96-301-00MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 96-301-00MG Supplier MilliporeSigma™ Supplier No. 9630100MG

missing translation for 'validMainframeMsg'

Chromogenic substrate used in conjunction with IPTG

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Color White
Quantity 100 mg
Solubility Information Water/DMF/IPTG/: 20mg/mL soluble
Formula Weight 408.63
Percent Purity ≥99.0%
Physical Form Powder
Chemical Name or Material X-Gal
Grade Molecular biology grade
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.