accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72420-38-3

IUPAC Name: 5-methyl-4-oxo-5-phenyl-4,5-dihydrofuran-2-carboxylic acid
Synonyms: acifran 阿昔呋喃 acifranum reductol
Molecular Weight (g/mol): 218.21
Molecular Formula: C12H10O4
InChi Key: DFDGRKNOFOJBAJ-UHFFFAOYNA-N
SMILES: CC1(OC(=CC1=O)C(O)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results