accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOFFFFFFNONHSNNFFF(E)

CAS RN 725247-18-7

IUPAC Name: (2E)-3-(4-{[2,4-bis(trifluoromethyl)phenyl]methoxy}-3-methoxyphenyl)-2-cyano-N-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]prop-2-enamide
Synonyms: (2E)-3-(4-{[2,4-bis(trifluoromethyl)phenyl]methoxy}-3-methoxyphenyl)-2-cyano-N-[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]prop-2-enamide
Molecular Weight (g/mol): 596.43
Molecular Formula: C23H13F9N4O3S
InChi Key: HQFNFOOGGLSBBT-AWNIVKPZSA-N
SMILES: COC1=CC(\C=C(/C#N)C(=O)NC2=NN=C(S2)C(F)(F)F)=CC=C1OCC1=CC=C(C=C1C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results