accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOHNNHOOCH3CH3H3C

CAS RN 72648-80-7

IUPAC Name: ethyl 2-[(2-{[(tert-butoxy)carbonyl]amino}ethyl)amino]acetate
Synonyms: ethyl 2-({2-[(tert-butoxycarbonyl)amino]ethyl}amino)acetate
Molecular Weight (g/mol): 246.31
Molecular Formula: C11H22N2O4
InChi Key: WVFXXOLKYIPCAX-UHFFFAOYSA-N
SMILES: CCOC(=O)CNCCNC(=O)OC(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results