CAS RN 72786-12-0
IUPAC Name:
hydrogen {[(2R,4S,7R)-6,11-dimethyl-6,11-diazatetracyclo[7.6.1.0²,⁷.0¹²,¹⁶]hexadeca-1(16),9,12,14-tetraen-4-yl]sulfamoyl}dimethylamine chloride
Synonyms:
mesulergine
Molecular Weight (g/mol):
398.95
Molecular Formula:
C18H27ClN4O2S
InChi Key:
HANSYUJEPWNHIM-IVMONYBCSA-N
SMILES:
[H+].[Cl-].CN(C)S(=O)(=O)N[C@H]1C[C@H]2[C@@H](CC3=CN(C)C4=CC=CC2=C34)N(C)C1
1
–
1
of
1
results
1
R&D Systems™ Mesulergine hydrochloride
CAS: 72786-12-0 Molecular Formula: C18H27ClN4O2S