CAS RN 72795-01-8
IUPAC Name:
1-[(7-methyl-2,3-dihydro-1H-inden-4-yl)oxy]-3-[(propan-2-yl)amino]butan-2-ol hydrochloride
Synonyms:
pertuzumab hydrochloride
Molecular Weight (g/mol):
313.87
Molecular Formula:
C17H28ClNO2
InChi Key:
KBXMBGWSOLBOQM-UHFFFAOYNA-N
SMILES:
Cl.CC(C)NC(C)C(O)COC1=C2CCCC2=C(C)C=C1
1
–
1
of
1
results
1
R&D Systems™ ICI 118,551 hydrochloride
CAS: 72795-01-8 Molecular Formula: C17H28ClNO2