accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 72797-16-1

IUPAC Name: 2-ethoxy-3-(propan-2-yl)pyrazine
Synonyms: 2-ethoxy-3-isopropylpyrazine
Molecular Weight (g/mol): 166.22
Molecular Formula: C9H14N2O
InChi Key: LTAUBPVQMBOANV-UHFFFAOYSA-N
SMILES: CCOC1=NC=CN=C1C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results