Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Ethoxy-3-isopropylpyrazine 95.0+%, TCI America™
SDP

Catalog No. E07165G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07165G Supplier TCI America Supplier No. E07165G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 72797-16-1
Molecular Formula C9H14N2O
Molecular Weight (g/mol) 166.22
MDL Number MFCD00038029
InChI Key LTAUBPVQMBOANV-UHFFFAOYSA-N
PubChem CID 175170
IUPAC Name 2-ethoxy-3-(propan-2-yl)pyrazine
SMILES CCOC1=NC=CN=C1C(C)C

Specifications

Color Yellow
Quantity 5 g
Formula Weight 166.22
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethoxy-3-isopropylpyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.