accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7295-76-3

IUPAC Name: 3-methoxypyridine
Synonyms: 3-methyoxypyridine
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: UMJSCPRVCHMLSP-UHFFFAOYSA-N
SMILES: COC1=CC=CN=C1
Boiling Point 
  • (2)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (4)