CAS RN 7298-99-9
IUPAC Name:
(2S,3S)-2-amino-3-hydroxybutanedioic acid
Synonyms:
L-threo-3-hydroxyaspartate threo-3-hydroxy-L-aspartate
Molecular Weight (g/mol):
149.1
Molecular Formula:
C4H7NO5
InChi Key:
YYLQUHNPNCGKJQ-LWMBPPNESA-N
SMILES:
N[C@@H]([C@H](O)C(O)=O)C(O)=O
1
–
1
of
1
results
1
R&D Systems™ L-(-)-threo-3-Hydroxyaspartic acid
CAS: 7298-99-9 Molecular Formula: C4H7NO5