accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7307-55-3

IUPAC Name: undecan-1-amine
Synonyms: N-undecylamin
Molecular Weight (g/mol): 171.33
Molecular Formula: C11H25N
InChi Key: QFKMMXYLAPZKIB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCN
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)