Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Undecylamine, MP Biomedicals

Catalog No. ICN219783 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219783 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219783 Supplier MP Biomedicals, Inc Supplier No. 0521978301

Chemical Identifiers

CAS 7307-55-3
Molecular Formula C11H25N
Molecular Weight (g/mol) 171.33
MDL Number MFCD00008152
InChI Key QFKMMXYLAPZKIB-UHFFFAOYSA-N
Synonym undecylamine, n-undecylamine, 1-aminoundecane, 1-undecanamine, hendecylamine, 1-undecylamine, monoundecylamine, n-undecylamin, unii-uze26a4ko1, uze26a4ko1
PubChem CID 81742
IUPAC Name undecan-1-amine
SMILES CCCCCCCCCCCN

Specifications

Quantity 10 g
Formula Weight 171.33
Chemical Name or Material n-Undecylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.