accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OHH3CH3CCH3CH3H3CCH3

CAS RN 732-26-3

IUPAC Name: 2,4,6-tri-tert-butylphenol
Synonyms: 2,4,6-tri(tert-butyl)phenol
Molecular Weight (g/mol): 262.44
Molecular Formula: C18H30O
InChi Key: PFEFOYRSMXVNEL-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results