CAS RN 7320-37-8
IUPAC Name:
2-tetradecyloxirane
Synonyms:
1,2-epoxyhexadecane hexadecene epoxide 1,2-hexadecane oxide 1,2-hexadecene epoxide
Molecular Weight (g/mol):
240.43
Molecular Formula:
C16H32O
InChi Key:
DSZTYVZOIUIIGA-UHFFFAOYNA-N
SMILES:
CCCCCCCCCCCCCCC1CO1
1
–
1
of
1
results
1
1,2-Epoxyhexadecane 80.0+%, TCI America™
CAS: 7320-37-8 Molecular Formula: C16H32O Molecular Weight (g/mol): 240.43 MDL Number: MFCD00005151 InChI Key: DSZTYVZOIUIIGA-UHFFFAOYNA-N Synonym: 1,2-Hexadecylene Oxide PubChem CID: 23741 ChEBI: CHEBI:34054 IUPAC Name: 2-tetradecyloxirane SMILES: CCCCCCCCCCCCCCC1CO1