CAS RN 73210-73-8
IUPAC Name:
(2E)-but-2-enedioic acid; bis(N-(2-{[2-hydroxy-3-(4-hydroxyphenoxy)propyl]amino}ethyl)morpholine-4-carboxamide)
Synonyms:
fumaric acid; bis(xamoterol)
Molecular Weight (g/mol):
794.86
Molecular Formula:
C36H54N6O14
InChi Key:
QEDVGROSOZBGOZ-WXXKFALUNA-N
SMILES:
OC(=O)\C=C\C(O)=O.OC(CNCCNC(=O)N1CCOCC1)COC1=CC=C(O)C=C1.OC(CNCCNC(=O)N1CCOCC1)COC1=CC=C(O)C=C1
1
–
1
of
1
results
1
R&D Systems™ Xamoterol hemifumarate
CAS: 73210-73-8 Molecular Formula: C36H54N6O14