accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3H3CCH3H3C

CAS RN 733-07-3

IUPAC Name: 1,3,5-trimethyl-2-[(2,4,6-trimethylphenyl)methyl]benzene
Synonyms: 1,3,5-trimethyl-2-[(2,4,6-trimethylphenyl)methyl]benzene
Molecular Weight (g/mol): 252.4
Molecular Formula: C19H24
InChi Key: ZQYQPPLKPFBKLD-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CC2=C(C)C=C(C)C=C2C)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results