accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 73513-42-5

IUPAC Name: 2-methylpentane
Synonyms: 2-methylpentane
Molecular Weight (g/mol): 86.18
Molecular Formula: C6H14
InChi Key: AFABGHUZZDYHJO-UHFFFAOYSA-N
SMILES: CCCC(C)C
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (4)
Grade 
  • (2)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)