CAS RN 73604-30-5
IUPAC Name:
6,6-dimethyl-5-{12-oxo-3,5,11-trioxatricyclo[7.3.0.0²,⁶]dodeca-1(9),2(6),7-trien-10-yl}-2H,5H,6H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium bromide
Synonyms:
6,6-dimethyl-5-{12-oxo-3,5,11-trioxatricyclo[7.3.0.0²,⁶]dodeca-1(9),2(6),7-trien-10-yl}-2H,5H,7H,8H-[1,3]dioxolo[4,5-g]isoquinolin-6-ium bromide
Molecular Weight (g/mol):
462.3
Molecular Formula:
C21H20BrNO6
InChi Key:
BWXCECYGGMGBHD-UHFFFAOYNA-M
SMILES:
[Br-].C[N+]1(C)CCC2=CC3=C(OCO3)C=C2C1C1OC(=O)C2=C1C=CC1=C2OCO1
1
–
1
of
1
results
1
R&D Systems™ (-)-Bicuculline methobromide
CAS: 73604-30-5 Molecular Formula: C21H20BrNO6