accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrOOBrNOHOOHONOOHOOH

CAS RN 73630-11-2

IUPAC Name: 2-{[2-(2-{2-[bis(carboxymethyl)amino]-5-bromophenoxy}ethoxy)-4-bromophenyl](carboxymethyl)amino}acetic acid
Synonyms: dibromo-bapta 5,5'-dibromo-BAPTA Br2-BAPTA
Molecular Weight (g/mol): 634.23
Molecular Formula: C22H22Br2N2O10
InChi Key: KCTRRYZOCJDOTC-UHFFFAOYSA-N
SMILES: OC(=O)CN(CC(O)=O)C1=C(OCCOC2=C(C=CC(Br)=C2)N(CC(O)=O)CC(O)=O)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results