accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7382-30-1

IUPAC Name: 2,5,8,11-tetraoxapentadecane
Synonyms: 2,5,8,11-tetraoxapentadecane
Molecular Weight (g/mol): 220.31
Molecular Formula: C11H24O4
InChi Key: SNAQINZKMQFYFV-UHFFFAOYSA-N
SMILES: CCCCOCCOCCOCCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results