Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylene Glycol Butyl Methyl Ether 98.0+%, TCI America™
SDP

Catalog No. T3181500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T3181500ML 500 mL
T318125ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T3181500ML Supplier TCI America Supplier No. T3181500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7382-30-1
Molecular Formula C11H24O4
Molecular Weight (g/mol) 220.31
MDL Number MFCD23703159
InChI Key SNAQINZKMQFYFV-UHFFFAOYSA-N
Synonym 1-[2-[2-(2-Methoxyethoxy)ethoxy]ethoxy]butane
PubChem CID 18344639
IUPAC Name 2,5,8,11-tetraoxapentadecane
SMILES CCCCOCCOCCOCCOC

Specifications

Melting Point -34°C
Color Colorless
Quantity 500 mL
Formula Weight 220.31
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Triethylene Glycol Butyl Methyl Ether
EINECSNumber (7)-1321
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.