accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 7382-32-3

IUPAC Name: 1-(2-butoxyethoxy)-2-methoxyethane
Synonyms: 1-(2-butoxyethoxy)-2-methoxyethane
Molecular Weight (g/mol): 176.26
Molecular Formula: C9H20O3
InChi Key: HYLLZXPMJRMUHH-UHFFFAOYSA-N
SMILES: CCCCOCCOCCOC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results