accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7382-37-8

IUPAC Name: 1-(methoxymethoxy)naphthalene
Synonyms: 1-(methoxymethoxy)naphthalene
Molecular Weight (g/mol): 188.23
Molecular Formula: C12H12O2
InChi Key: REELDIWIMZYNLC-UHFFFAOYSA-N
SMILES: COCOC1=C2C=CC=CC2=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results