Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Methoxymethoxy)naphthalene 97.0+%, TCI America™
SDP

Catalog No. M14545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M14545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M14545G Supplier TCI America Supplier No. M14545G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7382-37-8
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD06797119
InChI Key REELDIWIMZYNLC-UHFFFAOYSA-N
PubChem CID 10779684
IUPAC Name 1-(methoxymethoxy)naphthalene
SMILES COCOC1=CC=CC2=CC=CC=C21

Specifications

Color Yellow
Boiling Point 296°C
Quantity 5 g
Formula Weight 188.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Methoxymethoxy)naphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.