accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHHC

CAS RN 74-86-2

IUPAC Name: acetylene
Synonyms: acetylene ethine ethin narcylen vinylene acetylene (substance) azetylen
Molecular Weight (g/mol): 26.04
Molecular Formula: C2H2
InChi Key: HSFWRNGVRCDJHI-UHFFFAOYSA-N
SMILES: C#C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results