accessibility menu, dialog, popup

UserName

Viewing profile as user:
NClNCl

CAS RN 74065-64-8

IUPAC Name: 6,6'-bis(chloromethyl)-2,2'-bipyridine
Synonyms: 6,6'-bis(chloromethyl)-2,2'-bipyridine
Molecular Weight (g/mol): 253.13
Molecular Formula: C12H10Cl2N2
InChi Key: CUWSIBMCPYEBPL-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(=N1)C1=NC(CCl)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results