accessibility menu, dialog, popup

UserName

Viewing profile as user:
NBrHBr

CAS RN 74129-11-6

IUPAC Name: 4-bromopyridine hydrobromide
Synonyms: 4-bromopyridine hydrobromide
Molecular Weight (g/mol): 238.91
Molecular Formula: C5H5Br2N
InChi Key: SMSVBXDZZQOTGB-UHFFFAOYSA-N
SMILES: Br.BrC1=CC=NC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results