accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 741677-68-9

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium; trifluoro(trifluoromethyl)boranuide
Synonyms: 1-butyl-3-methylimidazolium; trifluoro(trifluoromethyl)boranuide
Molecular Weight (g/mol): 276.03
Molecular Formula: C9H15BF6N2
InChi Key: PTBVLMHYOBXNCB-UHFFFAOYSA-N
SMILES: F[B-](F)(F)C(F)(F)F.CCCCN1C=C[N+](C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results