accessibility menu, dialog, popup

UserName

Viewing profile as user:
N(R)NO(S)(R)NNNH2HOOPOPOPOOOOOHOOHNaNa

CAS RN 74299-50-6

IUPAC Name: disodium hydrogen [({[(2R,3S,5R)-5-(6-amino-9H-purin-9-yl)-3-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl phosphonato)oxy]phosphonate
Synonyms: disodium hydrogen {[(2R,3S,5R)-5-(6-aminopurin-9-yl)-3-hydroxyoxolan-2-yl]methoxy(hydroxy)phosphoryl phosphonato}oxyphosphonate
Molecular Weight (g/mol): 535.15
Molecular Formula: C10H14N5Na2O12P3
InChi Key: JEKDCIBJADJZSK-DZVPOJRENA-L
SMILES: [Na+].[Na+].NC1=C2N=CN([C@H]3C[C@H](O)[C@@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3)C2=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results