accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74328-56-6

IUPAC Name: 2-[2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-8-(oxiran-2-yl)octyl]oxirane
Synonyms: 2-[2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoro-8-(oxiran-2-yl)octyl]oxirane
Molecular Weight (g/mol): 414.19
Molecular Formula: C12H10F12O2
InChi Key: BUCVATDLDBZNPM-UHFFFAOYNA-N
SMILES: FC(F)(CC1CO1)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CC1CO1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results