accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7440-03-1

IUPAC Name: niobium
Synonyms: niobium|columbium|columbio|niobio|Niob
Molecular Weight (g/mol): 92.91
Molecular Formula: Nb
InChi Key: GUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES: [Nb]
Grade 
  • (1)
  • (4)
  • (2)
Solvent or Matrix 
  • (4)
Technique 
  • (4)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (4)
  • (2)
  • (4)
  • (4)
  • (4)