Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium ICP Standard, 1000 ppm Nb in 3% HNO3, Ricca Chemical
SDP

Catalog No. PNB1KN500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PNB1KN500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PNB1KN500 Supplier Ricca Chemical Company Supplier No. PNB1KN500
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7440-03-1 , 7697-37-2 , 7440-03-1 , 7664-39-3
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Packaging Natural Poly Bottle
CAS Min % 93.0, 4.89, 0.09, 0.09
CAS Max % 97.0, 5.09, 0.1, 0.1
Odor Odorless
Color Colorless
pH < 2
Boiling Point Approximately 100°C
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solubility Miscible with water
Solution Type Calibration Standard
Type Standard
Quantity 500 mL
Grade Laboratory
Concentration or Composition (by Analyte or Components) 1mL = 1mg Nb (1, 000ppm Nb)Nb in 3% HNO3 / trace HF
Chemical Name or Material Niobium ICP Standard
Show More Show Less
ShelfLife 18 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.