Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Niobium Standard, 1000μg/mL in Water with Trace Hydrofluoric Acid, Ultra Scientific

Catalog No. USICP041
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICP041 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICP041 Supplier Agilent Technologies Supplier No. ICP041
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7440-03-1 , 7664-39-3 , 7732-18-5
Molecular Formula Nb
Molecular Weight (g/mol) 92.91
MDL Number MFCD00011126
InChI Key GUCVJGMIXFAOAE-UHFFFAOYSA-N
PubChem CID 23936
ChEBI CHEBI:33344
IUPAC Name niobium
SMILES [Nb]

Specifications

Physical Form Liquid
Color Colorless
Quantity 125 mL
Chemical Name or Material Niobium Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.