accessibility menu, dialog, popup

UserName

Viewing profile as user:
Rb

CAS RN 7440-17-7

IUPAC Name: rubidium
Synonyms: rubidium|rubidium atom|rubidio
Molecular Weight (g/mol): 85.47
Molecular Formula: Rb
InChi Key: IGLNJRXAVVLDKE-UHFFFAOYSA-N
SMILES: [Rb]
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
Form 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (6)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)