CAS RN 7440-39-3
Barium crystalline dendritic solid, 99.9% (metals basis)
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N Synonym: bario,baryum,bario spanish,baryum french,barium, soluble compounds,unii-24gp945v5t,and soluble compounds,barium, elemental,and compounds,atom PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium rod, 22mm (0.9in) dia, 99+% (metals basis), Sr ∣<0.8%
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N Synonym: bario,baryum,bario spanish,baryum french,barium, soluble compounds,unii-24gp945v5t,and soluble compounds,barium, elemental,and compounds,atom PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium, 99+%, rods, approx. 2 cm diameter
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N Synonym: bario,baryum,bario spanish,baryum french,barium, soluble compounds,unii-24gp945v5t,and soluble compounds,barium, elemental,and compounds,atom PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium pieces, 2.5cm (1in) & down, 99.2% (metals basis), Sr ∣<0.8%
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N Synonym: bario,baryum,bario spanish,baryum french,barium, soluble compounds,unii-24gp945v5t,and soluble compounds,barium, elemental,and compounds,atom PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Tuning Solution 2 For ICP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Barium Standard For ICP/MS, Claritas PPT Grade, SPEX CertiPrep™
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium Standard in Base Oil, 5000μg/g, SPEX CertiPrep™
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Tuning Solution 3 For ICP, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Barium Standard For ICP, 10,000μg/mL in 5% HNO3, SPEX CertiPrep™
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Barium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
CAS: 7440-39-3 Molecular Formula: Ba Molecular Weight (g/mol): 137.33 MDL Number: MFCD00134031 InChI Key: DSAJWYNOEDNPEQ-UHFFFAOYSA-N PubChem CID: 5355457 ChEBI: CHEBI:32595 IUPAC Name: barium SMILES: [Ba]
Sigma Aldrich (S)-(+)-3-Acetyl-4-Benzyl-2-Oxazolidinone
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Barium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Barium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Barium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
- 1
- 2