CAS RN 7440-56-4
Germanium pieces, 3-9mm (0.1-0.4in), 99.999% (metals basis)
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N Synonym: element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium powder, -100 mesh, 99.999% (metals basis)
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N Synonym: element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium pieces, 2cm (0.8in) and down, Puratronic™, 99.999% (metals basis)
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N Synonym: element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium, plasma standard solution, Specpure™ Ge 1000μg/mL
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium pieces, 2cm (0.8in) & down, 99.9999% (metals basis)
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N Synonym: element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium, 99.999%, (trace metal basis), powder
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N Synonym: element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium rod, polycrystalline, 6.35mm (0.25in) dia, 99.9999% (metals basis)
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N Synonym: element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium pieces, 3.2mm (0.125in) and down, 99.999% (metals basis)
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N Synonym: element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N Synonym: element,germanio,germanium, metal powder,unii-00072j7xws,powder,germanium, chips trace metals basis,atom,copper metal,germanium, disks, 15mm, thickness 1.0mm, polycrystalline,germanium, powder PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Germanium Standard For ICP, 1000μg/mL in H2O, Assurance Grade, SPEX CertiPrep™
CAS: 7440-56-4 Molecular Formula: Ge Molecular Weight (g/mol): 72.63 MDL Number: MFCD00085310 InChI Key: GNPVGFCGXDBREM-UHFFFAOYSA-N PubChem CID: 6326954 ChEBI: CHEBI:30441 IUPAC Name: germanium SMILES: [Ge]
Sigma Aldrich Germanium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Germanium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Germanium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Germanium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Germanium
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.