accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3SiCH3CH3H2C

CAS RN 74472-22-3

IUPAC Name: tert-butyldimethyl(prop-2-en-1-yl)silane
Synonyms: tert-butyldimethylprop-2-en-1-ylsilane
Molecular Weight (g/mol): 156.34
Molecular Formula: C9H20Si
InChi Key: KALUNQGXJIFEDG-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)CC=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results