accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3NCO

CAS RN 745784-08-1

IUPAC Name: 1-(1-isocyanatoethyl)-3-methoxybenzene
Synonyms: 1-(1-isocyanatoethyl)-3-methoxybenzene
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: YNWCQMWZIOMQOP-UHFFFAOYNA-N
SMILES: COC1=CC=CC(=C1)C(C)N=C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results