accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 7459-33-8

IUPAC Name: (9E,12E)-octadeca-9,12-dienoyl chloride
Synonyms: (9E,12E)-octadeca-9,12-dienoyl chloride
Molecular Weight (g/mol): 298.9
Molecular Formula: C18H31ClO
InChi Key: FBWMYSQUTZRHAT-AVQMFFATSA-N
SMILES: CCCCC\C=C\C\C=C\CCCCCCCC(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results