accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74602-32-7

IUPAC Name: dodeca-5,7-diyne-1,12-diol
Synonyms: dodeca-5,7-diyne-1,12-diol
Molecular Weight (g/mol): 194.27
Molecular Formula: C12H18O2
InChi Key: SEWYHOMCDKWYEF-UHFFFAOYSA-N
SMILES: OCCCCC#CC#CCCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results