accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNCH3OCl

CAS RN 7461-38-3

IUPAC Name: 4-chloro-N,N-diethylbenzamide
Synonyms: 4-chloro-N,N-diethylbenzamide
Molecular Weight (g/mol): 211.69
Molecular Formula: C11H14ClNO
InChi Key: VQIFBGNOSBKNJI-UHFFFAOYSA-N
SMILES: CCN(CC)C(=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results