CAS RN 7462-74-0
IUPAC Name:
2-bromo-2-methylpropanamide
Synonyms:
2-bromo-2-methylpropanamide
Molecular Weight (g/mol):
166.02
Molecular Formula:
C4H8BrNO
InChi Key:
WTRIMJTZOOLIFZ-UHFFFAOYSA-N
SMILES:
CC(C)(Br)C(N)=O
1
–
1
of
1
results
1
2-Bromoisobutyramide 98.0+%, TCI America™
CAS: 7462-74-0 Molecular Formula: C4H8BrNO Molecular Weight (g/mol): 166.018 MDL Number: MFCD08234837 InChI Key: WTRIMJTZOOLIFZ-UHFFFAOYSA-N Synonym: 2-Bromo-2-methylpropionamide PubChem CID: 101235 IUPAC Name: 2-bromo-2-methylpropanamide SMILES: CC(C)(C(=O)N)Br