accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 74684-34-7

CH3ONHNNOH3COCH3OCH3OCH3OCH3OH3CNHOH3C

CAS RN 74684-34-7

IUPAC Name: 2,7,12,17-tetrakis(3,5-dimethoxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Synonyms: 2,7,12,17-tetrakis(3,5-dimethoxyphenyl)-21,22,23,24-tetraazapentacyclo[16.2.1.1³,⁶.1⁸,¹¹.1¹³,¹⁶]tetracosa-1,3,5,7,9,11,13(22),14,16,18(21),19-undecaene
Molecular Weight (g/mol): 854.96
Molecular Formula: C52H46N4O8
InChi Key: MOOXLDFVIJGBRF-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(OC)=C1)C1=C2NC(C=C2)=C(C2=NC(C=C2)=C(C2=NC(C=C2)=C(C2=CC=C1N2)C1=CC(OC)=CC(OC)=C1)C1=CC(OC)=CC(OC)=C1)C1=CC(OC)=CC(OC)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results