accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHOCH3NHNClOHSOHOO

CAS RN 747-36-4

IUPAC Name: 2-({4-[(7-chloroquinolin-4-yl)amino]pentyl}(ethyl)amino)ethan-1-ol; sulfuric acid
Synonyms: hydroxychloroquine; sulfuric acid
Molecular Weight (g/mol): 433.95
Molecular Formula: C18H28ClN3O5S
InChi Key: JCBIVZZPXRZKTI-UHFFFAOYNA-N
SMILES: OS(O)(=O)=O.CCN(CCO)CCCC(C)NC1=C2C=CC(Cl)=CC2=NC=C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (1)
  • (1)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)