accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)(R)CH3PPPhPhPhPh

CAS RN 74839-84-2

IUPAC Name: [(2R,3R)-3-(diphenylphosphanyl)butan-2-yl]diphenylphosphane
Synonyms: (R,R)-chiraphos
Molecular Weight (g/mol): 426.48
Molecular Formula: C28H28P2
InChi Key: FWXAUDSWDBGCMN-ZBXWWWQKNA-N
SMILES: C[C@H]([C@@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)