Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(R,R)-Chiraphos 97%, Thermo Scientific™

Catalog No. AC292422500
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC292422500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC292422500 Supplier Thermo Scientific Chemicals Supplier No. 292422500

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74839-84-2
Molecular Formula C28H28P2
Molecular Weight (g/mol) 426.48
MDL Number MFCD00075403
InChI Key FWXAUDSWDBGCMN-ZBXWWWQKNA-N
Synonym r,r-chiraphos, chiraphos, r,r, unii-1t086b9q1j, 2r,3r-+-bis diphenylphosphino butane, 2r,3r-butane-2,3-diylbis diphenylphosphine, 2r,3r-+-2,3-bis diphenylphosphino butane, cira, phosphine, 1r,2r-1,2-dimethyl-1,2-ethanediyl bis diphenyl, 2r,3r-bis diphenylphosphino butane, r,?r ?-?chiraphos
PubChem CID 2724948
IUPAC Name [(2R,3R)-3-(diphenylphosphanyl)butan-2-yl]diphenylphosphane
SMILES C[C@H]([C@@H](C)P(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 104°C to 109°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97%
Quantity 250 mg
Specific Rotation +188° to +204° (20°C, 589nm) (c=1.5, CHCl3)
Formula Weight 426.47
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material (R, R)-Chiraphos, 97%
Show More Show Less
Hazard Category AIR SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.