accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClCl

CAS RN 75-43-4

IUPAC Name: dichloro(fluoro)methane
Synonyms: dichlorofluoromethane
Molecular Weight (g/mol): 102.92
Molecular Formula: CHCl2F
InChi Key: UMNKXPULIDJLSU-UHFFFAOYSA-N
SMILES: FC(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results