accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFCl

CAS RN 75-72-9

IUPAC Name: chlorotrifluoromethane
Synonyms: chlorotrifluoromethane
Molecular Weight (g/mol): 104.46
Molecular Formula: CClF3
InChi Key: AFYPFACVUDMOHA-UHFFFAOYSA-N
SMILES: FC(F)(F)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results