Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorotrifluoromethane Solution, 100μg/mL in Methanol, Ultra Scientific

Catalog No. USCFC1201
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USCFC1201 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USCFC1201 Supplier Agilent Technologies Supplier No. CFC1201
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 75-72-9 , 67-56-1
Molecular Formula CClF3
Molecular Weight (g/mol) 104.456
InChI Key AFYPFACVUDMOHA-UHFFFAOYSA-N
PubChem CID 6392
IUPAC Name chloro(trifluoro)methane
SMILES C(F)(F)(F)Cl

Specifications

Physical Form Liquid
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Chlorotrifluoromethane, 100μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.